accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH(R)ONH2OH

CAS RN 153-94-6

IUPAC Name: (2R)-2-amino-3-(1H-indol-3-yl)propanoic acid
Synonyms: D-tryptophan|D-trytophane
Molecular Weight (g/mol): 204.23
Molecular Formula: C11H12N2O2
InChi Key: QIVBCDIJIAJPQS-SECBINFHSA-N
SMILES: N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O
Percent Purity 
  • (2)
  • (1)
  • (7)
Quantity 
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (7)
  • (2)
  • (1)
  • (7)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)