accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1530-03-6

IUPAC Name: (1-phenylpropyl)benzene
Synonyms: 1,1-diphenylpropane
Molecular Weight (g/mol): 196.29
Molecular Formula: C15H16
InChi Key: BUZMJVBOGDBMGI-UHFFFAOYSA-N
SMILES: CCC(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results