CAS RN 1530-12-7
IUPAC Name:
9H,9'H-9,9'-bifluorene
Synonyms:
9H,9'H-9,9'-bifluorene
Molecular Weight (g/mol):
330.43
Molecular Formula:
C26H18
InChi Key:
QXAIDADUIMVTPS-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(=C1)C(C1C3=CC=CC=C3C3=CC=CC=C13)C1=CC=CC=C21
1
–
1
of
1
results
1
9,9'-Bifluorenyl 98.0+%, TCI America™
CAS: 1530-12-7 Molecular Formula: C26H18 Molecular Weight (g/mol): 330.43 MDL Number: MFCD00001132 InChI Key: QXAIDADUIMVTPS-UHFFFAOYSA-N PubChem CID: 137063 IUPAC Name: 9-(9H-fluoren-9-yl)-9H-fluorene SMILES: C1=CC=C2C(=C1)C(C3=CC=CC=C32)C4C5=CC=CC=C5C6=CC=CC=C46