accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNIO

CAS RN 153034-90-3

IUPAC Name: 2-chloro-4-iodopyridine-3-carbaldehyde
Synonyms: 2-chloro-4-iodopyridine-3-carbaldehyde
Molecular Weight (g/mol): 267.45
Molecular Formula: C6H3ClINO
InChi Key: OPZCXONVRBYGHX-UHFFFAOYSA-N
SMILES: ClC1=NC=CC(I)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results