accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15305-72-3

IUPAC Name: hexaaminorutheniumbis(ylium) chloride
Synonyms: hexaaminorutheniumbis(ylium) chloride
Molecular Weight (g/mol): 232.66
Molecular Formula: ClH12N6Ru
InChi Key: MHYZHXDLILNYTQ-UHFFFAOYSA-M
SMILES: [Cl-].N[Ru++](N)(N)(N)(N)N
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)