accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15336-72-8

IUPAC Name: 4,4'-bipiperidine
Synonyms: 4,4'-bipiperidine
Molecular Weight (g/mol): 168.28
Molecular Formula: C10H20N2
InChi Key: PRNRUOJLUPUJDN-UHFFFAOYSA-N
SMILES: C1CC(CCN1)C1CCNCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results