CAS RN 15341-08-9
CAS RN 15341-08-9
IUPAC Name:
(5-nitro-2H-1,3-benzodioxol-4-yl)methanol
Synonyms:
(5-nitro-2H-1,3-benzodioxol-4-yl)methanol
Molecular Weight (g/mol):
197.15
Molecular Formula:
C8H7NO5
InChi Key:
LJXBRPHBDWIHPR-UHFFFAOYSA-N
SMILES:
OCC1=C2OCOC2=CC=C1[N+]([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals 6-Nitropiperonyl alcohol, 98+%
CAS: 15341-08-9 Molecular Formula: C8H7NO5 Molecular Weight (g/mol): 197.15 MDL Number: MFCD00005825 InChI Key: LJXBRPHBDWIHPR-UHFFFAOYSA-N Synonym: 6-nitropiperonyl alcohol,6-nitrobenzo d 1,3 dioxol-5-yl methanol,3,4-methylenedioxy-6-nitrobenzyl alcohol,6-nitro-3,4-methylenedioxybenzyl alcohol,6-nitro-1,3-benzodioxol-5-yl methanol,6-nitro-2h-1,3-benzodioxol-5-yl methanol,6-nitro-1,3-benzodioxole-5-methanol,1,3-benzodioxole-5-methanol,6-nitro,1,3-benzodioxole-5-methanol, 6-nitro,6-nitro-1,3-benzodioxol-5-yl methanol # PubChem CID: 519111 IUPAC Name: (5-nitro-2H-1,3-benzodioxol-4-yl)methanol SMILES: OCC1=C2OCOC2=CC=C1[N+]([O-])=O