accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CFOH

CAS RN 1534369-41-9

IUPAC Name: 3-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Synonyms: 3-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular Weight (g/mol): 238.07
Molecular Formula: C12H16BFO3
InChi Key: GXNDNRWVVOKEBG-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(O)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results