accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 153473-94-0

IUPAC Name: 1-(6,7-dimethoxyphthalazin-1-yl)piperidin-4-yl N-ethylcarbamate; 2-hydroxypropane-1,2,3-tricarboxylic acid
Synonyms: 1-(6,7-dimethoxyphthalazin-1-yl)piperidin-4-yl N-ethylcarbamate; citric acid
Molecular Weight (g/mol): 552.54
Molecular Formula: C24H32N4O11
InChi Key: QKMRPDVEKBYWPF-UHFFFAOYSA-N
SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O.CCNC(=O)OC1CCN(CC1)C1=C2C=C(OC)C(OC)=CC2=CN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results