accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3COH2NHCl

CAS RN 153502-27-3

IUPAC Name: O-[2-(trimethylsilyl)ethyl]hydroxylamine hydrochloride
Synonyms: O-[2-(trimethylsilyl)ethyl]hydroxylamine hydrochloride
Molecular Weight (g/mol): 169.72
Molecular Formula: C5H16ClNOSi
InChi Key: ZDNNDTUAVSQNGB-UHFFFAOYSA-N
SMILES: Cl.C[Si](C)(C)CCON
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results