accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSBr

CAS RN 153561-74-1

IUPAC Name: 2-bromo-5-dodecylthiophene
Synonyms: 2-bromo-5-dodecylthiophene
Molecular Weight (g/mol): 331.36
Molecular Formula: C16H27BrS
InChi Key: MOSAVYMGWTYDNS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CC=C(Br)S1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results