accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 153624-38-5

IUPAC Name: [4-(2-methylpropyl)phenyl]boronic acid
Synonyms: 4-(2-methylpropyl)phenylboronic acid
Molecular Weight (g/mol): 178.04
Molecular Formula: C10H15BO2
InChi Key: YZSPHVWALUJQNK-UHFFFAOYSA-N
SMILES: CC(C)CC1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results