CAS RN 153645-26-2
IUPAC Name:
tert-butyl N-(1-hydroxy-3,3-dimethylbutan-2-yl)carbamate
Synonyms:
tert-butyl N-(1-hydroxy-3,3-dimethylbutan-2-yl)carbamate
Molecular Weight (g/mol):
217.31
Molecular Formula:
C11H23NO3
InChi Key:
AZHJHZWFJVBGIL-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CO)C(C)(C)C
1
–
1
of
1
results
1
Sigma Aldrich ™-(-)-4-Penten-2-Ol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.