CAS RN 15366-08-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
116.87
Molecular Formula:
C4H9ClMg
InChi Key:
YNLPNVNWHDKDMN-UHFFFAOYSA-M
SMILES:
[Mg++].[Cl-].CC[CH-]C
1
–
1
of
1
results
1
Sigma Aldrich (S)-(-)-2-Amino-3-Phenyl-1-Propanol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.