CAS RN 153775-33-8
IUPAC Name:
2,4-difluoro-5-nitrobenzoate
Synonyms:
2,4-difluoro-5-nitrobenzoate
Molecular Weight (g/mol):
202.09
Molecular Formula:
C7H2F2NO4
InChi Key:
MDFSGDMPHMKKGB-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC(=C(F)C=C1F)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
2,4-Difluoro-5-nitrobenzoic acid, 97%
2,4-Difluoro-5-nitrobenzoic acid, 97%
CAS: 153775-33-8 Molecular Formula: C7H2F2NO4 Molecular Weight (g/mol): 202.09 MDL Number: MFCD04115713 InChI Key: MDFSGDMPHMKKGB-UHFFFAOYSA-M PubChem CID: 2782822 IUPAC Name: 2,4-difluoro-5-nitrobenzoate SMILES: [O-]C(=O)C1=CC(=C(F)C=C1F)[N+]([O-])=O