accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 153815-60-2

OOHONHHN(S)

CAS RN 153815-60-2

IUPAC Name: (9H-fluoren-9-yl)methyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
Synonyms: 9H-fluoren-9-ylmethyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
Molecular Weight (g/mol): 412.49
Molecular Formula: C26H24N2O3
InChi Key: TYDUDYNHTCJBHI-SFHVURJKSA-N
SMILES: OC[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)