accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15383-49-0

OHOOHOOHOOOH

CAS RN 15383-49-0

IUPAC Name: cyclohexane-1,2,4,5-tetracarboxylic acid
Synonyms: cyclohexane-1,2,4,5-tetracarboxylic acid
Molecular Weight (g/mol): 260.2
Molecular Formula: C10H12O8
InChi Key: ZPAKUZKMGJJMAA-UHFFFAOYNA-N
SMILES: OC(=O)C1CC(C(CC1C(O)=O)C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results