accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 153863-35-5

IUPAC Name: (5-methyl-1-phenyl-1H-pyrazol-4-yl)methanol
Synonyms: (5-methyl-1-phenylpyrazol-4-yl)methanol
Molecular Weight (g/mol): 188.23
Molecular Formula: C11H12N2O
InChi Key: JTNWKZVHKLAHTN-UHFFFAOYSA-N
SMILES: CC1=C(CO)C=NN1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results