accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNN

CAS RN 1539-42-0

IUPAC Name: bis[(pyridin-2-yl)methyl]amine
Synonyms: dipicolylamine
Molecular Weight (g/mol): 199.26
Molecular Formula: C12H13N3
InChi Key: KXZQYLBVMZGIKC-UHFFFAOYSA-N
SMILES: C(NCC1=CC=CC=N1)C1=CC=CC=N1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)