accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOOOOO

CAS RN 15397-15-6

IUPAC Name: 3,4-bis(benzoyloxy)-5-[(benzoyloxy)methyl]-3-methyloxolan-2-yl benzoate
Synonyms: 3,4-bis(benzoyloxy)-5-[(benzoyloxy)methyl]-3-methyloxolan-2-yl benzoate
Molecular Weight (g/mol): 580.59
Molecular Formula: C34H28O9
InChi Key: QJZSLTLDMBDKOU-UHFFFAOYNA-N
SMILES: CC1(OC(=O)C2=CC=CC=C2)C(OC(=O)C2=CC=CC=C2)OC(COC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results