accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1540-29-0

IUPAC Name: ethyl 2-acetylhexanoate
Synonyms: ethyl 2-acetylhexanoate
Molecular Weight (g/mol): 186.25
Molecular Formula: C10H18O3
InChi Key: ZTOQBHVLCJERBS-UHFFFAOYNA-N
SMILES: CCCCC(C(C)=O)C(=O)OCC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results