accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1540-34-7

H3CCH3OCH3O

CAS RN 1540-34-7

IUPAC Name: 3-ethylpentane-2,4-dione
Synonyms: 2,4-pentanedione, 3-ethyl-
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: GUARKOVVHJSMRW-UHFFFAOYSA-N
SMILES: CCC(C(C)=O)C(C)=O
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)