CAS RN 1541200-53-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Phenyl-5H-pyrido[3,2-b]indole 98.0+%, TCI America™
CAS: 1541200-53-6 Molecular Formula: C17H12N2 Synonym: 9-Phenyl-delta-carboline