accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNHNNNHN

CAS RN 1542234-40-1

IUPAC Name: 2,6-bis(2H-1,2,3-triazol-4-yl)pyridine
Synonyms: 2,6-bis(2H-1,2,3-triazol-4-yl)pyridine
Molecular Weight (g/mol): 213.2
Molecular Formula: C9H7N7
InChi Key: GTUXTIISPOXPEF-UHFFFAOYSA-N
SMILES: N1N=CC(=N1)C1=CC=CC(=N1)C1=NNN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results