accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 154230-29-2

IUPAC Name: [(1E)-2-(4-chlorophenyl)ethenyl]boronic acid
Synonyms: (1E)-2-(4-chlorophenyl)ethenylboronic acid
Molecular Weight (g/mol): 182.41
Molecular Formula: C8H8BClO2
InChi Key: HWSDRAPTZRYXHN-AATRIKPKSA-N
SMILES: OB(O)\C=C\C1=CC=C(Cl)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results