accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1544-68-9

IUPAC Name: 1-fluoro-4-isothiocyanatobenzene
Synonyms: 1-fluoro-4-isothiocyanatobenzene
Molecular Weight (g/mol): 153.17
Molecular Formula: C7H4FNS
InChi Key: NFIUJHJMCQQYDL-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)N=C=S
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results