accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1544-85-0

IUPAC Name: 2,2-difluoro-2H-1,3-benzodioxol-5-amine
Synonyms: 2,2-difluoro-1,3-benzodioxol-5-amine
Molecular Weight (g/mol): 173.12
Molecular Formula: C7H5F2NO2
InChi Key: CVYQRDKVWVBOFP-UHFFFAOYSA-N
SMILES: NC1=CC=C2OC(F)(F)OC2=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)