accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClPtH3CSH3CCH3SCH3

CAS RN 15442-57-6

IUPAC Name: bis((ethylsulfanyl)ethane); dichloroplatinumbis(ylium)
Synonyms: dichloroplatinumbis(ylium); bis(ethyl sulfide)
Molecular Weight (g/mol): 446.35
Molecular Formula: C8H20Cl2PtS2
InChi Key: RFSLRYGKHBDFGC-UHFFFAOYSA-L
SMILES: Cl[Pt++]Cl.CCSCC.CCSCC
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)