accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15442-91-8

IUPAC Name: 1,2,4,5-tetrakis(bromomethyl)benzene
Synonyms: 1,2,4,5-tetrakis(bromomethyl)benzene
Molecular Weight (g/mol): 449.81
Molecular Formula: C10H10Br4
InChi Key: UTXIKCCNBUIWPT-UHFFFAOYSA-N
SMILES: BrCC1=CC(CBr)=C(CBr)C=C1CBr
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)