accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNCl

CAS RN 154466-62-3

IUPAC Name: 2-chloro-5-pentylpyrimidine
Synonyms: 2-chloro-5-pentylpyrimidine
Molecular Weight (g/mol): 184.67
Molecular Formula: C9H13ClN2
InChi Key: WMARUIMXVALOMF-UHFFFAOYSA-N
SMILES: CCCCCC1=CN=C(Cl)N=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results