accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CFBr

CAS RN 1544739-25-4

IUPAC Name: 2-[3-(bromomethyl)-4-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[3-(bromomethyl)-4-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 314.99
Molecular Formula: C13H17BBrFO2
InChi Key: BQOMTZAOMQFSJP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(CBr)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results