accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHN

CAS RN 15450-05-2

IUPAC Name: 2-propyl-4,5-dihydro-1H-imidazole
Synonyms: 2-propyl-4,5-dihydro-1H-imidazole
Molecular Weight (g/mol): 112.18
Molecular Formula: C6H12N2
InChi Key: LFCYHAXVBWPGQR-UHFFFAOYSA-N
SMILES: CCCC1=NCCN1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results