accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHNNNH2H2O

CAS RN 15454-54-3

IUPAC Name: 2H-1,2,3,4-tetrazol-5-amine hydrate
Synonyms: 5-aminotetrazole hydrate
Molecular Weight (g/mol): 103.09
Molecular Formula: CH5N5O
InChi Key: JVSMPWHQUPKRNV-UHFFFAOYSA-N
SMILES: O.NC1=NNN=N1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)