accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 154549-38-9

IUPAC Name: [2,4,6-tris(propan-2-yl)phenyl]boronic acid
Synonyms: 2,4,6-triisopropylphenylboronic acid
Molecular Weight (g/mol): 248.17
Molecular Formula: C15H25BO2
InChi Key: FGFYEKLWZBTLEW-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(B(O)O)C(=C1)C(C)C
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)