accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15461-27-5

IUPAC Name: dichloro(oxo)zirconium
Synonyms: zirconium oxychloride|zirconyl chloride|ZrOCl2|zirconium chloride oxide|ZrCl2O|chlorozirconyl|zirconium oxydichloride
Molecular Weight (g/mol): 178.12
Molecular Formula: Cl2OZr
InChi Key: IPCAPQRVQMIMAN-UHFFFAOYSA-L
SMILES: Cl[Zr](Cl)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)