CAS RN 154674-81-4
IUPAC Name:
5-(2,6-dimethylbenzoyloxy)-3-(2-{2-[2-acetamido-3-(4-hydroxyphenyl)propanamido]-3-methylbutanamido}propanamido)-4-oxopentanoic acid
Synonyms:
5-(2,6-dimethylbenzoyloxy)-3-(2-{2-[2-acetamido-3-(4-hydroxyphenyl)propanamido]-3-methylbutanamido}propanamido)-4-oxopentanoic acid
Molecular Weight (g/mol):
654.72
Molecular Formula:
C33H42N4O10
InChi Key:
LOACDHZQOPEFKK-UHFFFAOYNA-N
SMILES:
CC(C)C(NC(=O)C(CC1=CC=C(O)C=C1)NC(C)=O)C(=O)NC(C)C(=O)NC(CC(O)=O)C(=O)COC(=O)C1=C(C)C=CC=C1C
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Caspase-1 Inhibitor IV
The Caspase-1 Inhibitor IV, also referenced under CAS 154674-81-4, controls the biological activity of Caspase-1. This small molecule/inhibitor is primarily used for Cancer applications.
2
Thermo Scientific Chemicals Caspase-1 Inhibitor IV
CAS: 154674-81-4