CAS RN 15469-77-9
IUPAC Name:
(3E)-dec-3-enoic acid
Synonyms:
(E)-3-decenoic acid dec-3t-enoic acid 3-decylenic acid trans-decen-3-saeure dec-3t-ensaeure
Molecular Weight (g/mol):
170.25
Molecular Formula:
C10H18O2
InChi Key:
CPVUNKGURQKKKX-BQYQJAHWSA-N
SMILES:
CCCCCC\C=C\CC(O)=O
1
–
1
of
1
results
1
3-Decenoic Acid 90.0+%, TCI America™
CAS: 15469-77-9 Molecular Formula: C10H18O2 Molecular Weight (g/mol): 170.252 MDL Number: MFCD00051464 InChI Key: CPVUNKGURQKKKX-BQYQJAHWSA-N PubChem CID: 5282725 ChEBI: CHEBI:37809 IUPAC Name: (E)-dec-3-enoic acid SMILES: CCCCCCC=CCC(=O)O