accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1547-87-1

IUPAC Name: 2,2,2-trifluoro-N,N-dimethylacetamide
Synonyms: 2,2,2-trifluoro-N,N-dimethylacetamide
Molecular Weight (g/mol): 141.09
Molecular Formula: C4H6F3NO
InChi Key: WXBWKMLIVXELSF-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results