accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1547491-70-2

IUPAC Name: 3-bromo-9,9-diphenyl-9H-fluorene
Synonyms: 3-bromo-9,9-diphenylfluorene
Molecular Weight (g/mol): 397.32
Molecular Formula: C25H17Br
InChi Key: AXVLLFWWMWDULG-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)C(C1=CC=CC=C21)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results