accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 154874-94-9

IUPAC Name: tert-butyl 4-chloropiperidine-1-carboxylate
Synonyms: tert-butyl 4-chloropiperidine-1-carboxylate
Molecular Weight (g/mol): 219.71
Molecular Formula: C10H18ClNO2
InChi Key: NZZWXABIGMMKQL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(Cl)CC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results