accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OO

CAS RN 154970-45-3

IUPAC Name: tert-butyl bicyclo[2.2.1]hept-5-ene-2-carboxylate
Synonyms: tert-butyl bicyclo[2.2.1]hept-5-ene-2-carboxylate
Molecular Weight (g/mol): 194.27
Molecular Formula: C12H18O2
InChi Key: BZBMBZJUNPMEBD-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C1CC2CC1C=C2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results