accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHO(S)O(R)OH(S)HO(S)OH(R)OHHO(E)

CAS RN 155-58-8

IUPAC Name: (2S,3R,4S,5S,6R)-2-{3-hydroxy-5-[(1E)-2-(3-hydroxy-4-methoxyphenyl)ethenyl]phenoxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: rhaponticin rhaponticine ponticin rhapontin (8CI) rhapontin
Molecular Weight (g/mol): 420.41
Molecular Formula: C21H24O9
InChi Key: GKAJCVFOJGXVIA-DXKBKAGUSA-N
SMILES: COC1=CC=C(\C=C\C2=CC(O)=CC(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)=C2)C=C1O
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)