accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15501-33-4

CH3CH3H3CI

CAS RN 15501-33-4

IUPAC Name: 1-iodo-2,2-dimethylpropane
Synonyms: 1-iodo-2,2-dimethylpropane
Molecular Weight (g/mol): 198.05
Molecular Formula: C5H11I
InChi Key: CJTZXIJETZZARD-UHFFFAOYSA-N
SMILES: CC(C)(C)CI
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results