accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOHOHOHOOHOOOOOOOOOHOOHOOOOOOOOOOHOHOOOOONNHNHNHNNHNHHNNH2NHHNHNNHH2NNHNHHNNH2NHH2NHNNHHNNH2NHNNHHNHNNHHNNHNHNH2H2NNH2(S)(S)(S)(S)(S)(S)(R)(S)(R)(R)H3CH3C(S)CH3(S)(S)(S)(S)H3CCH3(S)(R)(S)H3C(S)(S)CH3H3C(S)(S)(S)(S)CH3CH3(S)(S)CH3H3CH3CCH3

CAS RN 155069-90-2

IUPAC Name: (4S)-4-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-{[(1S,2R)-1-{[(2S)-1-[(2S)-2-{[(1S)-4-carbamimidamido-1-{[(1S,2R)-1-{[(1S)-2-carbamoyl-1-{[(1S,2R)-1-[({[(1S)-1-[({[(2S,3R)-1-[(2S)-2-carbamoylpyrrolidin-1-yl]-3-hydroxy-1-oxobutan-2-yl]carbamoyl}methyl)carbamoyl]-2-hydroxyethyl]carbamoyl}methyl)carbamoyl]-2-hydroxypropyl]carbamoyl}ethyl]carbamoyl}-2-hydroxypropyl]carbamoyl}butyl]carbamoyl}pyrrolidin-1-yl]-3-(4-hydroxyphenyl)-1-oxopropan-2-yl]carbamoyl}-2-hydroxypropyl]carbamoyl}-3-carbamoylpropyl]carbamoyl}-3-methylbutyl]carbamoyl}pentyl]carbamoyl}-2-(1H-imidazol-4-yl)ethyl]carbamoyl}-3-methylbutyl]carbamoyl}-4-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-6-amino-2-{2-[(2S)-2-[(2S)-2-amino-3-methylbutanamido]-4-methylpentanamido]acetamido}hexanamido]-4-methylpentanamido]-3-hydroxypropanamido]-4-carbamoylbutanamido]butanoic acid
Synonyms: (4S)-4-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-{[(1S,2R)-1-{[(2S)-1-[(2S)-2-{[(1S)-4-carbamimidamido-1-{[(1S,2R)-1-{[(1S)-2-carbamoyl-1-{[(1S,2R)-1-[({[(1S)-1-[({[(2S,3R)-1-[(2S)-2-carbamoylpyrrolidin-1-yl]-3-hydroxy-1-oxobutan-2-yl]carbamoyl}methyl)carbamoyl]-2-hydroxyethyl]carbamoyl}methyl)carbamoyl]-2-hydroxypropyl]carbamoyl}ethyl]carbamoyl}-2-hydroxypropyl]carbamoyl}butyl]carbamoyl}pyrrolidin-1-yl]-3-(4-hydroxyphenyl)-1-oxopropan-2-yl]carbamoyl}-2-hydroxypropyl]carbamoyl}-3-carbamoylpropyl]carbamoyl}-3-methylbutyl]carbamoyl}pentyl]carbamoyl}-2-(1H-imidazol-4-yl)ethyl]carbamoyl}-3-methylbutyl]carbamoyl}-4-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-6-amino-2-{2-[(2S)-2-[(2S)-2-amino-3-methylbutanamido]-4-methylpentanamido]acetamido}hexanamido]-4-methylpentanamido]-3-hydroxypropanamido]-4-carbamoylbutanamido]butanoic acid
Molecular Weight (g/mol): 2725.11
Molecular Formula: C119H198N36O37
InChi Key: CUQBOYYGZCRRKW-LPJWDPCBSA-N
SMILES: CC(C)C[C@H](NC(=O)[C@@H](N)C(C)C)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC1=CNC=N1)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC1=CC=C(O)C=C1)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H]([C@@H](C)O)C(=O)NCC(=O)N[C@@H](CO)C(=O)NCC(=O)N[C@@H]([C@@H](C)O)C(=O)N1CCC[C@H]1C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results