accessibility menu, dialog, popup

UserName

Viewing profile as user:
SSS

CAS RN 15509-95-2

IUPAC Name: 2-[3,5-bis(thiophen-2-yl)phenyl]thiophene
Synonyms: 2-[3,5-bis(thiophen-2-yl)phenyl]thiophene
Molecular Weight (g/mol): 324.47
Molecular Formula: C18H12S3
InChi Key: UBHPRZXDFVCNHZ-UHFFFAOYSA-N
SMILES: S1C=CC=C1C1=CC(=CC(=C1)C1=CC=CS1)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results