accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1551-21-9

IUPAC Name: 2-(methylsulfanyl)propane
Synonyms: 3-methyl-2-thiabutane
Molecular Weight (g/mol): 90.18
Molecular Formula: C4H10S
InChi Key: ROSSIHMZZJOVOU-UHFFFAOYSA-N
SMILES: CSC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results