accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrH3CP

CAS RN 15510-55-1

IUPAC Name: dodecyltriphenylphosphanium bromide
Synonyms: dodecyltriphenylphosphonium bromide
Molecular Weight (g/mol): 511.53
Molecular Formula: C30H40BrP
InChi Key: NSIFOGPAKNSGNW-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results