accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NClNOOCH3

CAS RN 15513-48-1

IUPAC Name: 4-chloro-2,6-dimethyl-3-nitropyridine
Synonyms: 4-chloro-2,6-dimethyl-3-nitropyridine
Molecular Weight (g/mol): 186.6
Molecular Formula: C7H7ClN2O2
InChi Key: PSBGFLWVQDGETK-UHFFFAOYSA-N
SMILES: CC1=NC(C)=C(C(Cl)=C1)[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)