accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO(E)(E)

CAS RN 1552-94-9

IUPAC Name: (2E,4E)-5-phenylpenta-2,4-dienoic acid
Synonyms: (2E,4E)-5-phenylpenta-2,4-dienoic acid
Molecular Weight (g/mol): 174.2
Molecular Formula: C11H10O2
InChi Key: FEIQOMCWGDNMHM-KBXRYBNXSA-N
SMILES: OC(=O)\C=C\C=C\C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results