accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15520-10-2

IUPAC Name: 2-methylpentane-1,5-diamine
Synonyms: 1,5-pentanediamine, 2-methyl-
Molecular Weight (g/mol): 116.21
Molecular Formula: C6H16N2
InChi Key: JZUHIOJYCPIVLQ-UHFFFAOYNA-N
SMILES: CC(CN)CCCN
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)